missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies SBI-0206965

Product Code. 17149991
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 17149991

Brand: STEMCELL Technologies 1000270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ULK1 and ULK2 kinase inhibitor

  • SBI-0206965 is an inhibitor of the autophagy initiator kinases ULK1 and ULK2 (IC50 = 108 nM and 711 nM, respectively; Egan et al.) SBI-0206965 is also a potent inhibitor of AMP-activated protein kinase (AMPK), a positive regulator of autophagy (Dite et al.)
  • Synergizes with mTOR inhibition to induce apoptosis and cell death in tumor cells (Egan et al.)
  • Suppresses autophagy induced by mTOR inhibition (Egan et al.)
  • Suppresses prosurvival autophagic responses in tumor cells through AMPK and ULK1 inhibition (Dite et al.)

Specifications

CAS 1884220-36-3
Quantity 10 mg
Molecular Formula C21H21BrN4O5
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.