missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies SBI-0206965
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
- SBI-0206965 is an inhibitor of the autophagy initiator kinases ULK1 and ULK2 (IC50 = 108 nM and 711 nM, respectively; Egan et al.) SBI-0206965 is also a potent inhibitor of AMP-activated protein kinase (AMPK), a positive regulator of autophagy (Dite et al.)
- Synergizes with mTOR inhibition to induce apoptosis and cell death in tumor cells (Egan et al.)
- Suppresses autophagy induced by mTOR inhibition (Egan et al.)
- Suppresses prosurvival autophagic responses in tumor cells through AMPK and ULK1 inhibition (Dite et al.)
Specifications
Specifications
| CAS | 1884220-36-3 |
| Quantity | 5 mg |
| Molecular Formula | C21H21BrN4O5 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction