missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SBF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576609
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Probable pseudophosphatase. Lacks several amino acids in the catalytic pocket which renders it catalytically inactive as a phosphatase. The pocket is however sufficiently preserved to bind phosphorylated substrates, and maybe protect them from phosphatases. Inhibits myoblast differentiation in vitro and induces oncogenic transformation in fibroblasts. According to PubMed:20937701, may function as a guanine nucleotide exchange factor (GEF) activating RAB28. Promotes the exchange of GDP to GTP, converting inactive GDP-bound Rab proteins into their active GTP-bound form.
Specifications
| SBF1 | |
| Polyclonal | |
| Unconjugated | |
| Sbf1 | |
| CMT4B3; DENN/MADD domain containing 7A; DENND7A; Inactive phosphatidylinositol 3-phosphatase 5; MTMR5; Myotubularin-related protein 5; Sbf1; SET binding factor 1; SET-binding factor 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 300147, 6305 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95248 | |
| Sbf1 | |
| Recombinant fusion protein of human SBF1. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction