missing translation for 'onlineSavingsMsg'
Learn More

SBE13 Hydrochloride, MedChemExpress

Product Code. 16442447
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442447 50 mg 50mg
16406338 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442447 Supplier Medchem Express Supplier No. HY15158

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SBE13 Hydrochloride is a potent and selective Plk1 inhibitor, with an IC50 of 200 pM; SBE13 Hydrochloride poorly inhibits Plk2 (IC50>66 μM) or Plk3 (IC50=875 nM).

SBE13 Hydrochloride is a potent and selective Plk1 inhibitor, with an IC50 of 200 pM

Specifications

Chemical Name or Material SBE13 Hydrochloride
CAS 1052532-15-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H28Cl2N2O4
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (208.59 mM) ∣H2O : 5 mg/mL (10.43 mM; Need ultrasonic)
SMILES COC1=CC=C(CCNCC2=CC=C(OCC3=CC=C(Cl)N=C3)C(OC)=C2)C=C1OC.[H]Cl
Molecular Weight (g/mol) 479.4
Formula Weight 479.4
Percent Purity 98.76%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.