missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SBE-β-CD, MedChemExpress
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
Unit Size:
10g
1g
25g
50g
5g
Description
SBE-β-CD is a sulfobutylether β-cyclodextrin derivative used as an excipient or a formulating agent to increase the solubility of poorly soluble agents.
Specifications
Specifications
| Chemical Name or Material | SBE-β-CD |
| CAS | 182410-00-0 |
| Color | White |
| Purity Grade Notes | Research |
| Quantity | 5 g |
| Synonym | Sulfobutylether-β-Cyclodextrin |
| Solubility Information | H2O : 125 mg/mL (Need ultrasonic) |
| SMILES | [R]O[C@@H]1[C@H](O[R])[C@H](O[C@@H]2[C@H](O[R])C(O[R])[C@H](O3)[C@@H](CO[R])O2)[C@@H](CO[R])O[C@@H]1O[C@H]4[C@H](O[R])[C@@H](O[R])[C@@H](O[C@H]5[C@H](O[R])[C@@H](O[R])[C@@H](O[C@@H]6C(O[R])[C@H](O[R])[C@H](O[C@@H]7[C@@H](O[R])[C@H](O[R])[C@H](O[C@@H]8[C@@H](O[R])[C@H](O[R])[C@H]3O[C@H]8CO[R])O[C@H]7CO[R])O[C@H]6CO[R])O[C@@H]5CO[R])O[C@@H]4CO[R].[R= H 21-m or C4H8SO3-Na+ m , m=6.0-7.1] |
| Grade | Research |
| Physical Form | Solid |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction