missing translation for 'onlineSavingsMsg'
Learn More

SBC-115076, MedChemExpress

Product Code. 16405148
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16405148 10 mg 10mg
16445856 100 mg 100mg
16427867 5 mg 5mg
16401377 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16405148 Supplier Medchem Express Supplier No. HY12402

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SBC-115076 is a potent proprotein convertase subtilisin/kexin type 9 (PCSK9) inhibitor. PCSK9 is a proprotein convertase, which plays a crucial role in LDL receptor metabolism.

SBC-115076 is a potent proprotein convertase subtilisin/kexin type 9 (PCSK9) inhibitor. PCSK9 is a proprotein convertase, which plays a crucial role in LDL receptor metabolism.

Specifications

Chemical Name or Material SBC-115076
CAS 489415-96-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C31H33N3O5
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (189.53 mM)
SMILES O=C1N(CCCN2CCOCC2)C(C3=CC=NC=C3)C(C(C4=CC=C(OCC5=CC=CC=C5)C(C)=C4)=O)=C1O
Molecular Weight (g/mol) 527.61
Formula Weight 527.61
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.