missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SB297006, MedChemExpress
Description
SB297006 is a CCR3 antagonist, which significantly inhibits proliferation and neurosphere formation in CCL11-treated neural progenitor cells.
Specifications
Specifications
| Chemical Name or Material | SB297006 |
| Physical Form | Solid |
| CAS | 58816-69-6 |
| Color | White |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C18H18N2O5 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : 150 mg/mL (438.15 mM; Need ultrasonic) |
| SMILES | O=C(OCC)[C@H](CC1=CC=C([N+]([O-])=O)C=C1)NC(C2=CC=CC=C2)=O |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?