missing translation for 'onlineSavingsMsg'
Learn More

SB290157 trifluoroacetate, MedChemExpress

Product Code. p-200099252
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16443398

Brand: Medchem Express HY101502A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SB290157 trifluoroacetate is a potent and selective C3a receptor antagonist with an IC50 of 200 nM.

SB290157 trifluoroacetate is a potent and selective C3a receptor antagonist with an IC50 of 200 nM.

Specifications

Chemical Name or Material SB290157 trifluoroacetate
Physical Form Powder
CAS 1140525-25-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H29F3N4O6
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 125 mg/mL (237.41 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (189.93 mM; Need ultrasonic)
SMILES N=C(N)NCCC[C@@H](C(O)=O)NC(COCC(C1=CC=CC=C1)C2=CC=CC=C2)=O.OC(C(F)(F)F)=O
Molecular Weight (g/mol) 526.51
Formula Weight 526.51
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.