missing translation for 'onlineSavingsMsg'
Learn More

SB290157 trifluoroacetate, MedChemExpress

Product Code. 16443398
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16443398 10 mg 10mg
16439926 50 mg 50mg
16406127 5 mg 5mg
16402756 25 mg 25mg
16424636 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16443398 Supplier Medchem Express Supplier No. HY101502A
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SB290157 trifluoroacetate is a potent and selective C3a receptor antagonist with an IC50 of 200 nM.

SB290157 trifluoroacetate is a potent and selective C3a receptor antagonist with an IC50 of 200 nM.

Specifications

Chemical Name or Material SB290157 trifluoroacetate
Physical Form Powder
CAS 1140525-25-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H29F3N4O6
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 125 mg/mL (237.41 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (189.93 mM; Need ultrasonic)
SMILES N=C(N)NCCC[C@@H](C(O)=O)NC(COCC(C1=CC=CC=C1)C2=CC=CC=C2)=O.OC(C(F)(F)F)=O
Molecular Weight (g/mol) 526.51
Formula Weight 526.51
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.