missing translation for 'onlineSavingsMsg'
Learn More

SB269652, MedChemExpress

Product Code. 16447817
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16447817 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16447817 Supplier Medchem Express Supplier No. HY12324
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SB269652 is the first drug-like allosteric modulator of the dopamine D2 receptor (D2R); a new chemical probe that can differentiate D2R monomers from dimers or oligomers depending on the observed pharmacology.

SB269652 is the first drug-like allosteric modulator of the dopamine D2 receptor (D2R)

Specifications

Chemical Name or Material SB269652
CAS 215802-15-6
Color White
Purity Grade Notes Research
Molecular Formula C27H30N4O
Quantity 5 mg
SMILES O=C(C1=CC2=C(C=CC=C2)N1)N[C@H]3CC[C@H](CCN(CC4)CC5=C4C=CC(C#N)=C5)CC3
Molecular Weight (g/mol) 426.55
Formula Weight 426.55
Percent Purity 98.95%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.