missing translation for 'onlineSavingsMsg'
Learn More

SB225002, MedChemExpress

Product Code. 16497146
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497146 100 mg 100mg
16493177 50 mg 50mg
16491226 200 mg 200mg
16449687 5 mg 5mg
16427158 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497146 Supplier Medchem Express Supplier No. HY16711
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SB225002, a potent, selective and non-peptide CXCR2 antagonist, inhibits 125I-IL-8 binding to CXCR2 with an IC50 of 22 nM.

SB225002, a potent, selective and non-peptide CXCR2 antagonist, inhibits 125I-IL-8 binding to CXCR2 with an IC50 of 22 nM.

Specifications

Chemical Name or Material SB225002
CAS 182498-32-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C13H10BrN3O4
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (283.98 mM)
SMILES O=C(NC1=CC=C([N+]([O-])=O)C=C1O)NC2=CC=CC=C2Br
Molecular Weight (g/mol) 352.14
Formula Weight 352.14
Percent Purity 99.77%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.