missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies SB216763
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
Description
- SB216763 is a cell-permeable ATP-competitive inhibitor of glycogen synthase kinase 3α (GSK3α, IC50 = 34 nM) and GSK3β isozymes (Coghlan et al.) GSK3 is a serine/threonine protein kinase that is inhibited by a variety of extracellular stimuli including insulin, growth factors, cell specification factors, and cell adhesion
Specifications
Specifications
| CAS | 280744-09-4 |
| Synonym | SB-216763 |
| Quantity | 5 mg |
| Molecular Formula | C19H12Cl2N2O2 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction