missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies SB202190

Product Code. p-7217629
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
This item is not returnable. View return policy

Product Code. 17100102

Brand: STEMCELL Technologies 72632

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

p38 MAPK inhibitor

  • SB202190 is a selective, potent, cell-permeable inhibitor of p38 MAP kinases, inhibiting p38α (SAPK2A, MAPK14) and p38β (SAPK2B, MAPK11) with IC50 values of 50 and 100 nM, respectively (Davies et al.; Jiang et al.) As a pyridinyl imidazole inhibitor, SB202190 directly binds p38 MAP kinases in the ATP binding pocket (Fox et al.)
  • Improves the self-renewal ability of neural stem cells from NPC1-deficient mice (Yang et al.)
  • Blocks adiponectin-mediated proliferation of hematopoietic stem cells (DiMascio et al.)
  • Reduces BMP3-mediated proliferation of C3H10T1/2 mesenchymal stem cells (Stewart et al.)
  • Induces cardiomyocyte differentiation from human embryonic stem cells (Graichen et al.)

Specifications

CAS 152121-30-7
Quantity 10 mg
Molecular Formula C20H14FN3O
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.