missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies SB202190

Product Code. 17110102
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17110102 25 mg 25mg
17100102 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17110102 Supplier STEMCELL Technologies Supplier No. 72634
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

p38 MAPK inhibitor

  • SB202190 is a selective, potent, cell-permeable inhibitor of p38 MAP kinases, inhibiting p38α (SAPK2A, MAPK14) and p38β (SAPK2B, MAPK11) with IC50 values of 50 and 100 nM, respectively (Davies et al.; Jiang et al.) As a pyridinyl imidazole inhibitor, SB202190 directly binds p38 MAP kinases in the ATP binding pocket (Fox et al.)
  • Improves the self-renewal ability of neural stem cells from NPC1-deficient mice (Yang et al.)
  • Blocks adiponectin-mediated proliferation of hematopoietic stem cells (DiMascio et al.)
  • Reduces BMP3-mediated proliferation of C3H10T1/2 mesenchymal stem cells (Stewart et al.)
  • Induces cardiomyocyte differentiation from human embryonic stem cells (Graichen et al.)

Specifications

CAS 152121-30-7
Quantity 25 mg
Molecular Formula C20H14FN3O
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.