missing translation for 'onlineSavingsMsg'
Learn More

SB-423562, MedChemExpress

Product Code. 16456318
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16456318 10 mg 10mg
16408907 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16456318 Supplier Medchem Express Supplier No. HY15105
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SB-423562 is a short-acting calcium-sensing receptor (CaR) antagonist. SB-423562 has the potential for osteoporosis research.

SB-423562 is a short-acting calcium-sensing receptor (CaR) antagonist. SB-423562 has the potential for osteoporosis research.

Specifications

Chemical Name or Material SB-423562
CAS 351490-27-2
Color White
Purity Grade Notes Research
Molecular Formula C26H32N2O4
Quantity 10 mg
Solubility Information DMSO : 5 mg/mL (11.45 mM; Need ultrasonic)
SMILES OC(CCC1=CC=C(C#N)C(OC[C@H](O)CNC(C)(C)CC2CC(C=CC=C3)=C3C2)=C1)=O
Molecular Weight (g/mol) 436.54
Formula Weight 436.54
Percent Purity 99.66%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.