missing translation for 'onlineSavingsMsg'
Learn More

SB 415286, MedChemExpress

Product Code. 16476518
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476518 10 mg 10mg
16432617 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16476518 Supplier Medchem Express Supplier No. HY15438
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SB 415286 is a potent and selective cell permeable inhibitor of GSK-3α, with an IC50 of 77.5 nM, and a Ki of 30.75 nM; SB 415286 is equally effective at inhibiting human GSK-3α and GSK-3β.

SB 415286 is a potent and selective cell permeable inhibitor of GSK-3α, with an IC50 of 77.5 nM, and a Ki of 30.75 nM

Specifications

Chemical Name or Material SB 415286
CAS 264218-23-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H10ClN3O5
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (277.99 mM; Need ultrasonic)
SMILES O=C(C(NC1=CC=C(O)C(Cl)=C1)=C2C3=CC=CC=C3[N+]([O-])=O)NC2=O
Molecular Weight (g/mol) 359.72
Formula Weight 359.72
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.