missing translation for 'onlineSavingsMsg'
Learn More

SB-334867 free base, MedChemExpress

Product Code. 16496967
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496967 5 mg 5mg
16474248 10 mg 10mg
16400587 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496967 Supplier Medchem Express Supplier No. HY10895A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SB-334867 free base (SB334867A free base) is an excellent, selective and blood-brain barrier permeable orexin-1 (OX1) receptor antagonist, shows selectivity over OX2 (pKb=7.4), 100-fold over 5-HT2B, 5-HT2C with pKi values of 5.4 and 5.3, respectively. SB-334867 reduces ethanol consumption and inhibits the acquisition of morphine-induced sensitization to locomotor activity in vivo.

SB-334867 free base (SB334867A free base) is an excellent, selective and blood-brain barrier permeable orexin-1 (OX1) receptor antagonist, shows selectivity over OX2 (pKb=7.4), 100-fold over 5-HT2B, 5-HT2C with pKi values of 5.4 and 5.3, respectively. SB-334867 reduces ethanol consumption and inhibits the acquisition of morphine-induced sensitization to locomotor activity in vivo.

Specifications

Chemical Name or Material SB-334867 free base
CAS 792173-99-0
Color White
Purity Grade Notes Research
Molecular Formula C17H13N5O2
Quantity 5 mg
Synonym SB334867A free base
Solubility Information DMSO : 50 mg/mL (156.58 mM; Need ultrasonic) ∣0.1 M HCL : 6 mg/mL (18.79 mM; ultrasonic and adjust pH to 3 with HCl)
SMILES O=C(NC1=CC(OC(C)=N2)=C2C=C1)NC3=CC=NC4=CC=CN=C43
Molecular Weight (g/mol) 319.32
Formula Weight 319.32
Percent Purity 99.33%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.