missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SB 290157 trifluoroacetate
GREENER_CHOICE

Product Code. 18735426
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18735426 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18735426 Supplier Tocris Bioscience™ Supplier No. 6860/5
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent C3a receptor antagonist

SB 290157 trifluoroacetate is a potent C3a receptor antagonist (IC50 values are 27 - 30 nM for C3a-induced Ca2+ mobilization in vitro and ex vivo). Inhibits C3a-induced chemotaxis in human mast cell line. Also inhibits tau hyperphosphorylation in vitro. Displays anti-inflammatory effects in animal models of airway inflammation and arthritis. Also inhibits glucose-induced insulin secretion and ATP production from ex vivo human and mouse pancreatic islets. Also exhibits C3a receptor agonist activity.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N2-[2-(2,2-Diphenylethoxy)acetyl]-L-arginine trifluoroacetate
CAS 1140525-25-2
Quantity 5 mg
Molecular Formula C22H28N4O4.CF3CO2H
Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.