missing translation for 'onlineSavingsMsg'
Learn More

SB-269970 hydrochloride, MedChemExpress

Product Code. 16422577
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16422577

Brand: Medchem Express HY15370A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SB-269970 hydrochloride is a potent, selective and brain-penetrant 5-HT7 receptor antagonist with a pKi of 8.3. SB-269970 hydrochloride exhibits >50-fold selectivity against other 5-HT receptors.

SB-269970 hydrochloride is a potent, selective and brain-penetrant 5-HT7 receptor antagonist with a pKi of 8.3. SB-269970 hydrochloride exhibits >50-fold selectivity against other 5-HT receptors.

Specifications

Chemical Name or Material SB-269970 hydrochloride
CAS 261901-57-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H29ClN2O3S
Quantity 50 mg
Synonym SB-269970A
Solubility Information DMSO : ≥ 30 mg/mL (77.13 mM) ∣H2O : 10 mg/mL (25.71 mM; ultrasonic and warming and heat to 60°C)
SMILES OC1=CC=CC(S(=O)(N2[C@@H](CCN3CCC(C)CC3)CCC2)=O)=C1.[H]Cl
Molecular Weight (g/mol) 388.95
Formula Weight 388.95
Percent Purity 98.62%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.