missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SB 258719 hydrochloride

Product Code. 18024342
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18024342 10 mg 10mg
18084752 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18024342 Supplier Tocris Bioscience™ Supplier No. 2726/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective 5-HT7 antagonist

Selective 5-HT7 receptor antagonist that displays > 100-fold selectivity over a range of other receptors. Reverses the hypothermic effect of 5-CT in mice following i.p. administration.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1217674-10-6
Molecular Formula C18H31ClN2O2S

Specifications

Quantity 10 mg
Formula Weight 374.97
Purity >99%
Molecular Weight (g/mol) 374.968
Chemical Name or Material SB 258719 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.