missing translation for 'onlineSavingsMsg'
Learn More

SB 242235, MedChemExpress

Product Code. 16497498
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497498 10 mg 10mg
16423527 50 mg 50mg
16419987 5 mg 5mg
16407466 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497498 Supplier Medchem Express Supplier No. HY18306

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SB-242235 is a potent and selective p38 MAP kinase inhibitor, with an IC50 of 1.0 μM in primary human chondrocytes.

SB-242235 is a potent and selective p38 MAP kinase inhibitor, with an IC50 of 1.0 μM in primary human chondrocytes.

Specifications

Chemical Name or Material SB 242235
CAS 193746-75-7
Color White
Purity Grade Notes Research
Molecular Formula C19H20FN5O
Quantity 10 mg
Solubility Information DMSO : ≥ 48 mg/mL (135.83 mM)
SMILES COC1=NC=CC(C2=C(C3=CC=C(F)C=C3)N=CN2C4CCNCC4)=N1
Molecular Weight (g/mol) 353.39
Formula Weight 353.39
Percent Purity 99.51%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.