missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SB 242084, TRC
Specifications
Specifications
| Chemical Name or Material | SB 242084 |
| CAS | 181632-25-7 |
| Molecular Formula | C21 H19 Cl N4 O2 . 2 Cl H |
| Quantity | 10 mg |
| Synonym | 6-Chloro-2,3-dihydro-5-methyl-N-[6-[(2-methyl-3-pyridinyl)oxy]-3-pyridinyl]-1H-indole-1-carboxamide |
| InChI Formula | InChI=1S/C21H19ClN4O2.2ClH/c1-13-10-15-7-9-26(18(15)11-17(13)22)21(27)25-16-5-6-20(24-12-16)28-19-4-3-8-23-14(19)2;;/h3-6,8,10-12H,7,9H2,1-2H3,(H,25,27);2*1H |
| SMILES | Cl.Cl.Cc1cc2CCN(C(=O)Nc3ccc(Oc4cccnc4C)nc3)c2cc1Cl |
| IUPAC Name | 6-chloro-5-methyl-N-[6-(2-methylpyridin-3-yl)oxypyridin-3-yl]-2,3-dihydroindole-1-carboxamide;dihydrochloride |
| Molecular Weight (g/mol) | 467.78 |
| Formula Weight | 466.073 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?