missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SB 242084
GREENER_CHOICE

Product Code. 30001369
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001369

Brand: Tocris Bioscience™ 2901/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective 5-HT2C antagonist; brain penetrant

5-HT2C receptor antagonist that displays 158- and 100-fold selectivity over 5-HT2A and 5-HT2B receptors respectively. Also displays selectivity over a range of other 5-HT, dopamine and adrenergic receptors. Brain penetrant; exerts anxiolytic-like activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1215566-78-1
Molecular Formula C21H21Cl3N4O2

Specifications

Quantity 10 mg
Formula Weight 467.78
Purity >99%
Molecular Weight (g/mol) 467.775
Chemical Name or Material SB 242084
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.