missing translation for 'onlineSavingsMsg'
Learn More

SB 242084 dihydrochloride, MedChemExpress

Product Code. 16465668
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465668 10 mg 10mg
16418367 5 mg 5mg
16411837 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16465668 Supplier Medchem Express Supplier No. HY13409A
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SB 242084 hydrochloride is a 5-HT2C receptor antagonist(pKi=9.0) that displays 158- and 100-fold selectivity over 5-HT2A and 5-HT2B receptors respectively.

SB 242084 hydrochloride is a 5-HT2C receptor antagonist(pKi=9.0) that displays 158- and 100-fold selectivity over 5-HT2A and 5-HT2B receptors respectively.

Specifications

Chemical Name or Material SB 242084 dihydrochloride
CAS 1049747-87-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H21Cl3N4O2
Quantity 10 mg
Solubility Information DMSO : ≥ 125.5 mg/mL (268.29 mM) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(N1CCC2=C1C=C(Cl)C(C)=C2)NC3=CC=C(OC4=CC=CN=C4C)N=C3.[H]Cl.[H]Cl
Molecular Weight (g/mol) 467.78
Formula Weight 467.78
Percent Purity 98.33%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.