missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SB 221284, Tocris Bioscience™
Potent, selective 5-HT2C/2B antagonist
Chemical Identifiers
CAS | 196965-14-7 |
---|---|
Molecular Formula | C16H14F3N3OS |
Molecular Weight (g/mol) | 353.363 |
InChI Key | OQZOXHCRSXYSPM-UHFFFAOYSA-N |
Synonym | tocris-1379, 2,3-dihydro-5-methylthio-n-3-pyridinyl-6-trifluoromethyl-1h-indole-1-carboxamide, 5-methylsulfanyl-n-pyridin-3-yl-6-trifluoromethyl-2,3-dihydroindole-1-carboxamide, d0dy5g, gtpl191, 5-methylthio-1-3-pyridylcarbamoyl-6-trifluoromethyl indoline, 5-methylthio-n-pyridin-3-yl-6-trifluoromethyl indoline-1-carboxamide, 5-methylsulfanyl-n-3-pyridyl-6-trifluoromethyl indoline-1-carboxamide |
PubChem CID | 443389 |
ChEBI | CHEBI:8978 |
IUPAC Name | 5-methylsulfanyl-N-pyridin-3-yl-6-(trifluoromethyl)-2,3-dihydroindole-1-carboxamide |
SMILES | CSC1=C(C=C2C(=C1)CCN2C(=O)NC3=CN=CC=C3)C(F)(F)F |
Description
Potent, selective 5-HT2C/2B receptor antagonist. pKi values are 6.4, 7.9 and 8.6 for 5-HT2A, 2B and 2C receptors respectively. Centrally active upon systemic administration in vivo.Chemical Identifiers
196965-14-7 | |
353.363 | |
tocris-1379, 2,3-dihydro-5-methylthio-n-3-pyridinyl-6-trifluoromethyl-1h-indole-1-carboxamide, 5-methylsulfanyl-n-pyridin-3-yl-6-trifluoromethyl-2,3-dihydroindole-1-carboxamide, d0dy5g, gtpl191, 5-methylthio-1-3-pyridylcarbamoyl-6-trifluoromethyl indoline, 5-methylthio-n-pyridin-3-yl-6-trifluoromethyl indoline-1-carboxamide, 5-methylsulfanyl-n-3-pyridyl-6-trifluoromethyl indoline-1-carboxamide | |
CHEBI:8978 | |
CSC1=C(C=C2C(=C1)CCN2C(=O)NC3=CN=CC=C3)C(F)(F)F |
C16H14F3N3OS | |
OQZOXHCRSXYSPM-UHFFFAOYSA-N | |
443389 | |
5-methylsulfanyl-N-pyridin-3-yl-6-(trifluoromethyl)-2,3-dihydroindole-1-carboxamide |
Specifications
196965-14-7 | |
tocris-1379, 2,3-dihydro-5-methylthio-n-3-pyridinyl-6-trifluoromethyl-1h-indole-1-carboxamide, 5-methylsulfanyl-n-pyridin-3-yl-6-trifluoromethyl-2,3-dihydroindole-1-carboxamide, d0dy5g, gtpl191, 5-methylthio-1-3-pyridylcarbamoyl-6-trifluoromethyl indoline, 5-methylthio-n-pyridin-3-yl-6-trifluoromethyl indoline-1-carboxamide, 5-methylsulfanyl-n-3-pyridyl-6-trifluoromethyl indoline-1-carboxamide | |
CSC1=C(C=C2C(=C1)CCN2C(=O)NC3=CN=CC=C3)C(F)(F)F | |
353.363 | |
CHEBI:8978 | |
>99% |
C16H14F3N3OS | |
OQZOXHCRSXYSPM-UHFFFAOYSA-N | |
5-methylsulfanyl-N-pyridin-3-yl-6-(trifluoromethyl)-2,3-dihydroindole-1-carboxamide | |
443389 | |
353.36 | |
SB 221284 |
Safety and Handling
Recommended Storage : Store at +4°C