missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SB 204990

Description
SB 204990 is an ATP citrate lyase (ACLY) inhibitor; it is a prodrug of SB 201076. SB 204990 inhibits cholesterol and fatty acid synthesis in a dose-dependent manner in HepG2 cells. Suppresses growth of cancer cells showing aerobic glycolysis in vitro and in vivo. Orally active.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | SB 204990 |
| Purity | >98% |
| Molecular Weight (g/mol) | 389.27 |
| Inhibitors | SB 204990 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?