missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SB 204990
GREENER_CHOICE

Product Code. 18730688
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18730688 10 mg 10mg
18767007 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18730688 Supplier Tocris Bioscience™ Supplier No. 4962/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ATP citrate lyase (ACLY) inhibitor

SB 204990 is an ATP citrate lyase (ACLY) inhibitor; it is a prodrug of SB 201076. SB 204990 inhibits cholesterol and fatty acid synthesis in a dose-dependent manner in HepG2 cells. Suppresses growth of cancer cells showing aerobic glycolysis in vitro and in vivo. Orally active.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type SB 204990
Purity >98%
Molecular Weight (g/mol) 389.27
Inhibitors SB 204990

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.