missing translation for 'onlineSavingsMsg'
Learn More

Saxagliptin, MedChemExpress

Product Code. 16463758
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16463758 10 mg 10mg
16400147 50 mg 50mg
16424826 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16463758 Supplier Medchem Express Supplier No. HY10285
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Saxagliptin (BMS-477118) is a potent, selective, reversible, competitive and orally active dipeptidyl peptidase-4 (DPP-4) (Ki = 0.6-1.3 nM) inhibitor. Saxagliptin has the peotential for type 2 diabetes mellitus research.

Saxagliptin (BMS-477118) is a potent, selective, reversible, competitive and orally active dipeptidyl peptidase-4 (DPP-4) (Ki = 0.6-1.3 nM) inhibitor. Saxagliptin has the peotential for type 2 diabetes mellitus research.

Specifications

Chemical Name or Material Saxagliptin
CAS 361442-04-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H25N3O2
Quantity 10 mg
Synonym BMS-477118
Solubility Information DMSO : ≥ 34 mg/mL (107.80 mM) ∣H2O : 10 mg/mL (31.70 mM; Need ultrasonic)
SMILES N[C@@H]([C@](C[C@H](C1)C2)(C[C@@H]1C3)C[C@]23O)C(N([C@@H](C4)C#N)[C@@H]5[C@H]4C5)=O
Molecular Weight (g/mol) 315.41
Formula Weight 315.41
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.