missing translation for 'onlineSavingsMsg'
Learn More

Savolitinib, MedChemExpress

Product Code. 16492987
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492987 50 mg 50mg
16479477 5 mg 5mg
16417016 100 mg 100mg
16416938 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16492987 Supplier Medchem Express Supplier No. HY15959
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Savolitinib (AZD-6094) is a potent, highly selective, and orally bioavailable c-Met inhibitor with IC50 s of 5 nM and 3 nM for c-Met and p-Met, respectively. Savolitinib (AZD-6094) selectively binds to and inhibits the activation of c-Met in an ATP-competitive manner, and disrupts c-Met signal transduction pathways. Antineoplastic activity.

Savolitinib (AZD-6094) is a potent, highly selective, and orally bioavailable c-Met inhibitor with IC50 s of 5 nM and 3 nM for c-Met and p-Met, respectively. Savolitinib (AZD-6094) selectively binds to and inhibits the activation of c-Met in an ATP-competitive manner, and disrupts c-Met signal transduction pathways. Antineoplastic activity.

Specifications

Chemical Name or Material Savolitinib
CAS 1313725-88-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H15N9
Quantity 50 mg
Synonym Volitinib HMPL-504 AZD-6094
Solubility Information DMSO : 25 mg/mL (72.39 mM; Need ultrasonic)
SMILES CN1N=CC(C2=CN=C3C(N([C@H](C4=CN5C(C=C4)=NC=C5)C)N=N3)=N2)=C1
Molecular Weight (g/mol) 345.36
Formula Weight 345.36
Percent Purity 98.45%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.