missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAV1 Antibody (3B2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00060485-M02
This item is not returnable.
View return policy
Description
SAV1 Monoclonal antibody specifically detects SAV1 in Human, Mouse, Yeast samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| SAV1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| 45 kDa WW domain protein, hWW45, protein salvador homolog 1, salvador homolog 1 (Drosophila), SAV, WW domain-containing, WW45salvador, WWP4,1700040G09Rik | |
| SAV1 (NP_068590, 300 a.a. ∽ 383 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. HTAEIPDWLQVYARAPVKYDHILKWELFQLADLDTYQGMLKLLFMKELEQIVKMYEAYRQALLTELENRKQRQQWYAQQHGKNF | |
| 0.1 mg | |
| Cell Biology, Signal Transduction | |
| 60485 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 3B2 | |
| Western Blot 1:500, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin | |
| NP_068590 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human, Mouse, Yeast | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction