missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Shamrock™ Satin Cloth Satin Cloth Labels
Click to view available options
Size (Metric):
25 x 57 mm
38 X 57 mm
Unit Size:
Pack of 1000
Specifications
Specifications
| Adhesive Type | Permanent |
| Color | White |
| Material | Satin Cloth |
| Product Type | Satin Cloth Label |
| For Use With (Application) | Cryogenic use |
| Size (Metric) | 38 X 57 mm |
| Dimensions (LxW) | 38 x 57 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction