missing translation for 'onlineSavingsMsg'
Learn More

Shamrock™ Satin Cloth Satin Cloth Labels

Product Code. 11740474
Click to view available options
Size (Metric):
25 x 57 mm
38 X 57 mm
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 11740474

Brand: Shamrock™ SC10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Adhesive Type Permanent
Color White
Material Satin Cloth
Product Type Satin Cloth Label
For Use With (Application) Cryogenic use
Size (Metric) 38 X 57 mm
Dimensions (LxW) 38 x 57 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.