missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SATB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119602
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
DNA-binding protein SATB2 binds to DNA at the nuclear matrix- or scaffold associated regions. It is thought to recognize the sugar-phosphate structure of double-stranded DNA. SATB2 is a transcription factor controlling nuclear gene expression by binding to matrix attachment regions (MARs) of DNA and inducing a local chromatin-loop remodeling. It also acts as a docking site for several chromatin remodeling enzymes by recruiting corepressors (HDACs) or coactivators (HATs) directly to promoters and enhancers. SATB2 is required for the initiation of the upper-layer neurons (UL1) specific genetic program and for the inactivation of deep-layer neurons (DL) and UL2 specific genes - probably by modulating BCL11B expression. It is a repressor of Ctip2 and a regulatory determinant of corticocortical connections in the developing cerebral cortex. Mutations in the gene are associate with cleft palate isolated (CPI).
Specifications
| SATB2 | |
| Polyclonal | |
| Unconjugated | |
| SATB2 | |
| DNA-binding protein SATB2; FLJ21474; FLJ32076; GLSS; KIAA1034; KIAA1034-like DNA binding protein; MGC119474; MGC119477; mKIAA1034; RGD1562369; SATB family member 2; SATB homeobox 2; SATB2; special AT-rich sequence binding protein 2; special AT-rich sequence-binding protein 2; two cut domains-containing homeodomain protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23314, 501145 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1.34 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9UPW6 | |
| SATB2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SATB2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction