missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SATB2 Polyclonal Antibody
Description
A suggested positive control is mouse brain tissue lysate. PA5-20532 can be used with blocking peptide PEP-0652.
DNA-binding protein SATB2 binds to DNA at the nuclear matrix- or scaffold associated regions. It is thought to recognize the sugar-phosphate structure of double-stranded DNA. SATB2 is a transcription factor controlling nuclear gene expression by binding to matrix attachment regions (MARs) of DNA and inducing a local chromatin-loop remodeling. It also acts as a docking site for several chromatin remodeling enzymes by recruiting corepressors (HDACs) or coactivators (HATs) directly to promoters and enhancers. SATB2 is required for the initiation of the upper-layer neurons (UL1) specific genetic program and for the inactivation of deep-layer neurons (DL) and UL2 specific genes - probably by modulating BCL11B expression. It is a repressor of Ctip2 and a regulatory determinant of corticocortical connections in the developing cerebral cortex. Mutations in the gene are associate with cleft palate isolated (CPI).
Specifications
Specifications
| Antigen | SATB2 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SATB2 |
| Gene Accession No. | Q8VI24, Q9UPW6 |
| Gene Alias | DNA-binding protein SATB2; FLJ21474; FLJ32076; GLSS; KIAA1034; KIAA1034-like DNA binding protein; MGC119474; MGC119477; mKIAA1034; RGD1562369; SATB family member 2; SATB homeobox 2; SATB2; special AT-rich sequence binding protein 2; special AT-rich sequence-binding protein 2; two cut domains-containing homeodomain protein |
| Gene Symbols | SATB2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?