missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SATB2 Antibody (rSATB2/8635), Novus Biologicals™
Mouse Monoclonal Antibody
£178.00 - £351.00
Specifications
| Antigen | SATB2 |
|---|---|
| Clone | rSATB2/8635 |
| Concentration | 0.2 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18606004
|
Novus Biologicals
NBP3-23797-20ug |
20 μg |
£178.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680535
|
Novus Biologicals
NBP3-23797-100ug |
100 μg |
£351.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SATB2 Monoclonal antibody specifically detects SATB2 in Human samples. It is validated for ELISASpecifications
| SATB2 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DNA-binding protein SATB2, FLJ32076, KIAA1034FLJ21474, MGC119474, MGC119477, SATB family member 2, SATB homeobox 2, Special AT-rich sequence-binding protein 2 | |
| Recombinant fragment (around aa150-350) of human SATB2 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| rSATB2/8635 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 23314 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title