missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SATB2 Antibody (CL0276), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52900
This item is not returnable.
View return policy
Description
SATB2 Monoclonal antibody specifically detects SATB2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SATB2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL0276 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| DNA-binding protein SATB2, FLJ32076, KIAA1034FLJ21474, MGC119474, MGC119477, SATB family member 2, SATB homeobox 2, Special AT-rich sequence-binding protein 2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VSQAVFARVAFNRTQGLLSEILRKEEDPRTASQSLLVNLRAMQNFLNLPEVERDRIYQDERERSMNPNVSMVSSASSSPSSSRTPQAKTSTPTTDLPIKVDGANINITAAIYDEIQQEMKRAK | |
| 0.1 mL | |
| Signal Transduction | |
| 23314 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction