missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SATB1 Polyclonal Antibody

Description
A suggested positive control is SK-N-SH cell lysate. PA5-20535 can be used with blocking peptide PEP-0655.
SATB1 (DNA-binding protein SATB1) is a crucial silencing factor that contributes to the initiation of X inactiviation mediated by Xist RNA that occurs during embryogenesis and in lymphoma. SATB1 binds to DNA at special AT-rich sequences, the consensus SATB1-binding sequence, and at nuclear matrix- or scaffold-associated regions. It is thought to recognize the sugar-phosphate structure of double-stranded DNA. SATB1 is also a transcriptonal repressor controlling nuclear and viral gene expression in a phosphorylated and acetylated status-dependent manner by binding to matrix attachment regions of DNA and inducing a local chromatin-loop remodeling. SATB1 modulates the genes that are essential in the maturatio of the immune T-cell CD8SP from thymocytes. Alu-like motifs and SATB1-binding sites provide an unique chromatin context which seems preferentially targeted by the HIV-1 integration machinery. Moreover, HIV-1 Tat may overcome SATB1-mediated repression of IL1 and IL2RA in T-cells by binding to the same domain as HDAC1.
Specifications
Specifications
| Antigen | SATB1 |
| Applications | ChIP Assay, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Satb1 |
| Gene Accession No. | Q01826, Q60611 |
| Gene Alias | 2610306G12Rik; AW413156; DNA-binding protein SATB1; SATB homeobox 1; Satb1; special AT-rich sequence binding protein 1; special AT-rich sequence binding protein 1 (binds to nuclear matrix/scaffold-associating DNA); special AT-rich sequence-binding protein 1 |
| Gene Symbols | Satb1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?