missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SATB1 Antibody (PCRP-SATB1-2C3), Novus Biologicals™
Mouse Monoclonal Antibody
£178.00 - £351.00
Specifications
| Antigen | SATB1 |
|---|---|
| Clone | PCRP-SATB1-2C3 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/Immunofluorescence 1-2 μg/mL |
| Applications | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043682
|
Novus Biologicals™
NBP3-26726-100ug |
100 μg |
£351.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043594
|
Novus Biologicals™
NBP3-26726-20ug |
20 μg |
£178.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SATB1 Monoclonal antibody specifically detects SATB1 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry/ ImmunofluorescenceSpecifications
| SATB1 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| DNA-binding protein SATB1, SATB homeobox 1, special AT-rich sequence binding protein 1 (binds to nuclearmatrix/scaffold-associating DNA's), Special AT-rich sequence-binding protein 1 | |
| Recombinant full-length human SATB1 protein | |
| Primary | |
| Store at 4°C. |
| PCRP-SATB1-2C3 | |
| Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/Immunofluorescence 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 6304 | |
| IgG | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title