missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554566
This item is not returnable.
View return policy
Description
Immunogen sequence: IYVMPEYRGQ GIGSKIIKKV AEVALDKGCS QFRLAVLDWN QRAMDLYKAL GAQDLTEAEG WHFFCFQGEA TRKLAGK Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.
SAT2 is an enzyme which catalyzes the acetylation of polyamines. Substrate specificity: norspermidine > spermidine = spermine >> N(1)acetylspermine = putrescine.
Specifications
| SAT2 | |
| Polyclonal | |
| Unconjugated | |
| Sat2 | |
| 2610016A03Rik; Diamine acetyltransferase 2; diamine N-acetyltransferase 2; EGK_08139; Polyamine N-acetyltransferase 2; Sat2; Spermidine/spermine N(1)-acetyltransferase 2; spermidine/spermine N1-acetyl transferase 2; spermidine/spermine N1-acetyltransferase family member 2; Ssat2; SSAT-2; thialysine N-epsilon-acetyltransferase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 112483 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96F10 | |
| Sat2 | |
| Recombinant protein corresponding to Human SAT2. Recombinant protein control fragment (Product #RP-93296). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Condividi una correzione di contenuto