missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAT2 Monoclonal Antibody (OTI2B3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525940
This item is not returnable.
View return policy
Description
SAT2 Monoclonal Antibody for Western Blot, IHC (P)
SAT2 is an enzyme which catalyzes the acetylation of polyamines. Substrate specificity: norspermidine > spermidine = spermine >> N(1)acetylspermine = putrescine.
Specifications
| SAT2 | |
| Monoclonal | |
| 0.58 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6P8J2, Q96F10 | |
| Sat2 | |
| Full length human recombinant protein of SAT2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B3 | |
| Unconjugated | |
| Sat2 | |
| 2610016A03Rik; Diamine acetyltransferase 2; diamine N-acetyltransferase 2; EGK_08139; Polyamine N-acetyltransferase 2; Sat2; Spermidine/spermine N(1)-acetyltransferase 2; spermidine/spermine N1-acetyl transferase 2; spermidine/spermine N1-acetyltransferase family member 2; Ssat2; SSAT-2; thialysine N-epsilon-acetyltransferase | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 112483, 69215 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction