missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10708-1-AP-150UL
This item is not returnable.
View return policy
Description
Spermidine/spermine-N1-acetyltransferase (SSAT1) is a short-lived polyamine catabolic enzyme which catalyzes the acetylation of polyamines. This highly regulated enzyme allows a fine attenuation of the intracellular concentration of polyamines. Induction of SSAT1 plays an important role in polyamine homoeostasis, since the N1-acetylated polyamines can be excreted or oxidized by acetylpolyamine oxidase.Specifications
| SAT1 | |
| Polyclonal | |
| Unconjugated | |
| SAT1 | |
| DC21, Diamine acetyltransferase 1, KFSD, Putrescine acetyltransferase, SAT, SAT1, SSAT, SSAT 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20229, 302642, 6303 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P21673, P48026 | |
| SAT1 | |
| SAT1 Fusion Protein Ag1154 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction