missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106434
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SAT1.
Spermidine/spermine-N1-acetyltransferase (SSAT1) is a short-lived polyamine catabolic enzyme which catalyzes the acetylation of polyamines. This highly regulated enzyme allows a fine attenuation of the intracellular concentration of polyamines. Induction of SSAT1 plays an important role in polyamine homoeostasis, since the N1-acetylated polyamines can be excreted or oxidized by acetylpolyamine oxidase.
Specifications
| SAT1 | |
| Polyclonal | |
| Unconjugated | |
| SAT1 | |
| AA617398; Ab2-402; CAON; DC21; diamine acetyltransferase 1; diamine acetyltransferase 1; spermidine/spermine N1-acetyltransferase 1; diamine N-acetyltransferase 1; diamine N-acetyltransferase 1 a; EC 2.3.1.57; EDM4; epid34; hypothetical protein LOC508861; kfsd; KFSDX; Polyamine N-acetyltransferase 1; putrescine acetyltransferase; SAT; Sat1; SAT-1; sat1.S; sat1a; sat1-a; sat1-b; Slc26a1; solute carrier family 26 (anion exchanger), member 1; solute carrier family 26 (sulfate transporter), member 1; solute carrier family 26 member 1; solute carrier family 26, member 1; Spermidine / spermine N1-acyltransferase (diamine acetyltransferase); spermidine/spermine N(1)-acetyltransferase 1; spermidine/spermine N1-acetyl transferase 1; spermidine/spermine N1-acetyltransferase; spermidine/spermine N1-acetyltransferase 1; spermidine/spermine N1-acetyltransferase 1 a; spermidine/spermine N1-acetyltransferase 1 S homeolog; spermidine/spermine N1-acetyltransferase alpha; spermidine/spermine N-acetyltransferase; SSAT; SSAT-1; sulfate anion tranporter AT1; sulfate anion transporter 1; sulfate anion transporter-1; sulfate/anion transporter SAT-1 protein; XELAEV_18014544mg | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20229, 302642, 6303 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P21673, P48026 | |
| SAT1 | |
| A synthesized peptide derived from human SAT1(Accession P21673), corresponding to amino acid residues C122-E171. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction