missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAT1 Mouse anti-Human, Clone: CPTC-SAT1-3, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07768-100ug
This item is not returnable.
View return policy
Description
SAT1 Monoclonal specifically detects SAT1 in Human samples. It is validated for ELISA, Protein Array.
Specifications
| SAT1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Protein Array | |
| DC21, diamine acetyltransferase 1, diamine N-acetyltransferase 1, EC 2.3.1.57, KFSD, Polyamine N-acetyltransferase 1, Putrescine acetyltransferase, SAT, Spermidine/spermine N(1)-acetyltransferase 1, spermidine/spermine N1-acetyltransferase, spermidine/spermine N1-acetyltransferase 1, spermidine/spermine N1-acetyltransferase alpha, SSAT-1, SSATKFSDX | |
| Recombinant full-length human SAT1 protein | |
| 100 μg | |
| Cancer, Cell Biology, Hypoxia, Mitochondrial Markers | |
| 6303 | |
| Store at 4°C. | |
| IgG2a κ |
| ELISA, Peptide Array | |
| CPTC-SAT1-3 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction