missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SASH1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15622535
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15622535 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15622535 Supplier Invitrogen™ Supplier No. PA548234

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SASH1 Polyclonal Antibody for Western Blot, IHC (P)

May have a role in a signaling pathway. Could act as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SASH1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SASH1
Gene Accession No. O94885, P59808
Gene Alias 1100001C18Rik; 2500002E12Rik; A330076K04Rik; dJ323M4; dJ323M4.1; KIAA0790; mKIAA0790; PEPE1; Proline-glutamate repeat-containing protein; putative adapter and scaffold protein 1; RGD1566017; SAM and SH3 domain containing 1; SAM and SH3 domain-containing protein 1; Sash1; SH3D6A
Gene Symbols SASH1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 596-625 amino acids from the Central region of human SASH1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23328, 70097
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.