missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SASH1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-26650V
This item is not returnable.
View return policy
Description
SASH1 Polyclonal antibody specifically detects SASH1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| SASH1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| dJ323M4, dJ323M4.1, KIAA0790RP3-323M4.1, PEPE1, Proline-glutamate repeat-containing protein, SAM and SH3 domain containing 1,2500002E12Rik, SAM and SH3 domain-containing protein 1, SH3D6A | |
| The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human SAM and SH3 domain containing 1 (NP_056093.3). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 23328 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction