missing translation for 'onlineSavingsMsg'
Learn More

SASH1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18403646 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403646 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403646 Supplier Novus Biologicals Supplier No. NBP126651AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SASH1 Polyclonal antibody specifically detects SASH1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SASH1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias dJ323M4, dJ323M4.1, KIAA0790RP3-323M4.1, PEPE1, Proline-glutamate repeat-containing protein, SAM and SH3 domain containing 1,2500002E12Rik, SAM and SH3 domain-containing protein 1, SH3D6A
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 750 and 800 of human SAM and SH3 domain containing 1 using the numbering given in entry NP_056093.3 (GeneID 23328).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23328
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.