missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane
BIOPHARMA

Product Code. 10062741 Shop All Sartorius Products
Change view
Click to view available options
Pore Size:
0.2 μm
Quantity:
100/Pk.
25/Pk.
25/Pk. (Starter Pack) 5 Ea.
Unit Size:
Pack of 100
Pack of 25
MWCO:
10 kDa
100 kDa
1000 kDa
3 kDa
30 kDa
300 kDa
5 kDa
50 kDa
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Quantity unitSize MWCO
10062741 0.2 μm 100/Pk. Pack of 100 -
10384931 - 25/Pk. Pack of 25 5 kDa
10083930 - 100/Pk. Pack of 100 10 kDa
10243281 - 25/Pk. Pack of 25 30 kDa
10191321 - 25/Pk. Pack of 25 10 kDa
10527251 - 100/Pk. Pack of 100 5 kDa
10551351 - 100/Pk. Pack of 100 30 kDa
10009450 - 25/Pk. Pack of 25 50 kDa
10561351 - 100/Pk. Pack of 100 50 kDa
10596651 - 25/Pk. Pack of 25 100 kDa
10245471 - 100/Pk. Pack of 100 100 kDa
11748813 - 25/Pk. Pack of 25 300 kDa
10609541 - 100/Pk. Pack of 100 1000 kDa
11730726 - 100/Pk. Pack of 100 300 kDa
10708402 - 25/Pk. Pack of 25 1000 kDa
10153711 0.2 μm 25/Pk. Pack of 25 -
10357791 - 25/Pk. Pack of 25 3 kDa
10302412 - 100/Pk. Pack of 100 3 kDa
10019780 - 25/Pk. (Starter Pack) 5 Ea. Pack of 25 300 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
This item is not returnable. View return policy
Product Code. 10062741 Supplier Sartorius Supplier No. VS0272

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Combines high speed with superior retentate recovery for concentrating very dilute solutions. Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane feature twin vertical membranes, small internal surface areas and thin-channel chambers. Available in a range of MWCO and packaging quantities.

Vivaspin concentrators are disposable ultrafiltration devices for concentrating biological samples. Vivaspin 2 is designed for use with samples from 500μl to 2mL in rotors that accept 15mL centrifuge tubes. The unique design provides high-speed concentrations for even particle-laden solutions and near-total concentrate recovery.

Twin vertical membranes and optimal design for fast, safe concentration

  • Patented twin vertical membrane design for ultra-fast processing speeds
  • In a single spin, solution volumes up to 4mL can be concentrated up to 30x in 5-30 minutes
  • Thin-channel filtration chamber minimizes membrane fouling and increases filtration speed
  • Dead stop pocket built into the bottom eliminates the risk of sample filtration to dryness
  • High-flux PES membranes feature very low protein binding and low extractables

Designed for maximum flexibility and sample recovery

  • Low internal surface and membrane area for superior recoveries from very dilute solutions
  • Two methods for easy concentrate recovery:
    • Directly pipet the concentrate from the dead stop pocket in the concentrator
    • Reverse-spin the concentrate into the recovery cap, which can be sealed for storage
    • Both methods provide near-total concentrate recovery of greater than 90%
  • Volume graduation for easy monitoring of concentrate volume and 8μl dead-stop volume
  • Universal compatibility with swing bucket or fixed angle rotors accepting 15mL centrifuge tubes
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) PES
Pore Size 0.2 μm
Sample Volume Up to 3 mL
Sterility Non-sterile
Filtration Area 1.3 cm2
For Use With (Application) Concentration, desalting and buffer applications
Housing Material Polycarbonate
Max. Force Swing bucket rotor: 4000 x g, Fixed angle rotor: 9000 x g (3 - 50 kDa MWCO); 12,000 x g (100 kDa+ MWCO)
Quantity 100/Pk.
Type Centrifugal Concentrator
Volume (Metric) 2 mL
Length (Metric) 126 mm
Width (Metric) 17.8 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.