missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Vivaspin™ 2 Centrifugal Concentrator with Cellulose Triacetate (CTA) Membrane

Product Code. 10190991 Shop All Sartorius Products
Change view
Click to view available options
MWCO:
10 kDa
20 kDa
5 kDa
Quantity:
100 Pk.
25 Pk.
Unit Size:
Pack of 100
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Quantity unitSize
10190991 5 kDa 25 Pk. Pack of 25
10140611 5 kDa 100 Pk. Pack of 100
11785785 10 kDa 25 Pk. Pack of 25
10448552 10 kDa 100 Pk. Pack of 100
11795785 20 kDa 25 Pk. Pack of 25
10101331 20 kDa 100 Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10190991 Supplier Sartorius™ Supplier No. VS02U1

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Vivaspin™ 2 Centrifugal Concentrators are disposable ultrafiltration units for concentration of macromolecules. With a broad range of molecular weight cut-offs (MWCO) and membrane materials they are suitable for various substrates, including proteins, viruses, nucleic acids, and nanoparticles.

Vivaspin concentrators are designed for the concentration or diafiltration of initial sample volumes up to 3 mL and can be used with swinging bucket or fixed angle rotors that accept 15 mL centrifuge tubes. Vivaspin's patented vertical membrane design and thin channel filtration chamber (US 5,647,990) minimize membrane fouling and provide high-speed concentration for even particle-laden solutions. The speed of Vivaspin products combined with low internal surface and membrane area allow superior recovery from very dilute solutions. The Vivaspin 2 also offers flexibility for process optimization: one can directly pipette the concentrate from the dead stop pocket (built into the bottom of the concentrator) or reverse-spin the concentrate into the recovery cap (which can be sealed for storage). Either method results in near-total concentrate recovery.

  • High retentate recovery >90%
  • Vertical membrane design for ultra fast concentration
  • Universal rotor compatibility
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Optional re-spin for concentrate recovery
  • Volume graduation for easy reading/monitoring
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Membrane Filter
For Use With (Equipment) Swinging Bucket Rotor (3 mL), Fixed-angle Rotor (2 mL)
Material (Membrane) Cellulose Triacetate
MWCO 5 kDa
Sample Volume 2 mL
Sterility Non-sterile
Quantity 25 Pk.
Type Centrifugal Concentrator
Volume (Metric) 2 mL
Length (English) 5 in.
Length (Metric) 126 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.