missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivapure™ S Mini H IEX Spin Columns
BIOPHARMA

Product Code. 11770855 Shop All Sartorius Products
Change view
Click to view available options
Capacity (Metric):
0.4 mL
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11770855 0.4 mL Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11770855 Supplier Sartorius Supplier No. VSIX01SH24
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ready-to-use centrifugal devices based on Sartobind Membrane Adsorber technology as their chromatography matrix

Vivapure IEX spin columns make protein purification as easy as filtration. The devices are ready-to-use and do not bear the risk of running dry. For many protein purification applications, they can replace time-consuming and tedious column chromatography. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many different samples are processed in parallel.

  • Fast centrifugal spin columns
  • Easy and ready-to-use
  • Ideal for parallel processing
  • Sample preparation prior to 1-D or 2-D PAGE
  • Purification scouting of an unknown protein
  • Removal of endotoxins from monoclonal antibodies
  • Preparation of heme moiety from heme containing protein for functional analysis
  • Polishing of His-tagged proteins
  • General protein purification
  • Detergent removal from protein solutions
  • Purification of antibodies from serum, ascites or tissue culture supernatant
  • Establishing a purification/ pre-purification protocol for a given protein
  • HPLC/FPLC sample preparation
  • Purification of membrane-bound proteins
TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 240 μL
Capacity (Metric) 0.4 mL
Material Polypropylene
Pore Size 3 to 5 μm
Support Type Stabilized regenerated Cellulose
Quantity 24/Pk.
For Use With (Application) Sample fractionation, purification condition scouting, small scale purification
Type Spin Column
Sample Volume 24 x 0.5 mL
Ligand Type Sulfonic Acid
Sufficient For Protein Purification
Column Type Ion Exchange Spin Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.