Learn More
Description
Vivapure IEX spin columns make protein purification as easy as filtration. The devices are ready-to-use and do not bear the risk of running dry. For many protein purification applications, they can replace time-consuming and tedious column chromatography. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many different samples are processed in parallel.
- Fast centrifugal spin columns
- Easy and ready-to-use
- Ideal for parallel processing
- Sample preparation prior to 1-D or 2-D PAGE
- Purification scouting of an unknown protein
- Removal of endotoxins from monoclonal antibodies
- Preparation of heme moiety from heme containing protein for functional analysis
- Polishing of His-tagged proteins
- General protein purification
- Detergent removal from protein solutions
- Purification of antibodies from serum, ascites or tissue culture supernatant
- Establishing a purification/ pre-purification protocol for a given protein
- HPLC/FPLC sample preparation
- Purification of membrane-bound proteins
Specifications
Specifications
| Bed Volume | 2700 μL |
| Capacity (Metric) | 19 mL |
| Material | Polypropylene |
| Pore Size | 3 to 5 μm |
| Support Type | Stabilized regenerated Cellulose |
| Quantity | 8/Pk. |
| For Use With (Application) | Large scale sample fractionation, one step Protein purification/concentration, polishing of His-tagged Protein |
| Type | Spin Column |
| Sample Volume | 8 x 20 mL |
| Ligand Type | Sulfonic Acid |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
